Alfa Aesar™ Benzyltrimethylammonium hydroxide, 20% w/w aq. soln.

Manufacturer: Alfa Aesar™ 41717AK

Quantity 250mL

 View more versions of this product

Catalog No. AA41717AK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100-85-6
    Linear Formula C6H5CH2N(CH3)3OH
    Formula Weight 167.25
    MDL Number MFCD00008281
    UN Number UN3267
    Chemical Name or Material Benzyltrimethylammonium hydroxide

    Specifications

    Formula Weight 167.25
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    UN Number UN3267
    Odor Amine-like
    Sensitivity Air Sensitive
    RTECS Number BO8575000
    Linear Formula C6H5CH2N(CH3)3OH
    MDL Number MFCD00008281
    CAS 100-85-6
    Color Colorless to pale yellow
    Concentration 20% w/w aq. soln.
    TSCA TSCA
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
    Quantity 250mL
    Chemical Name or Material Benzyltrimethylammonium hydroxide

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit