Learn More
Benzyltrimethylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals
Description
Benzyltrimethylammonium hydroxide is used as a phase-transfer catalyst. It is also used in aldol condensation reactions and base-catalyzed dehydration reactions. Further, it serves as a strong organic base and used in Horner-Wadsworth-Emmons olefination reactions. In addition, it is an active component in the formulation, which is used in silicon wafer cleaning applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless to Yellow |
Density | 1.059 g/mL |
Odor | Amine-like |
Refractive Index | 1.43 |
Linear Formula | C6H5CH2N(CH3)3OH |
Quantity | 100 mL |
UN Number | UN3267 |
Beilstein | 3917256 |
Sensitivity | Air Sensitive |
Solubility Information | Miscible with alcohols,hydrocarbons,aromatic hydrocarbons and halogenated solvents. |
Show More |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S. |
EINECSNumber | 202-895-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.