Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltrimethylammonium tribromide, 97%, Acros Organics

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
111865-47-5
C10H16Br3N
389.95
MFCD00134423
GSDDWZVWOXMLJZ-UHFFFAOYSA-N
benzyltrimethylammonium tribromide
86222689
benzyl(trimethyl)azanium;molecular bromine;hydrobromide
C[N+](C)(C)CC1=CC=CC=C1.Br.BrBr
111865-47-5
C10H16Br3N
389.95
MFCD00134423
GSDDWZVWOXMLJZ-UHFFFAOYSA-N
benzyltrimethylammonium tribromide
86222689
benzyl(trimethyl)azanium;molecular bromine;hydrobromide
C[N+](C)(C)CC1=CC=CC=C1.Br.BrBr
Melting Point 97°C to 101°C
Quantity 50g
Color Yellow to Orange
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (Iodometry)
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol
Formula Weight 389.95
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Benzyltrimethylammonium tribromide
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning