Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyltrimethylammonium tribromide, 98%

Click to view available options
Quantity:
1g
5g
25g
111865-47-5
C10H17Br3N+
390.965
MFCD00134423
GSDDWZVWOXMLJZ-UHFFFAOYSA-N
benzyltrimethylammonium tribromide
86222689
benzyl(trimethyl)azanium;molecular bromine;hydrobromide
C[N+](C)(C)CC1=CC=CC=C1.Br.BrBr
111865-47-5
C10H17Br3N+
390.965
MFCD00134423
GSDDWZVWOXMLJZ-UHFFFAOYSA-N
benzyltrimethylammonium tribromide
86222689
benzyl(trimethyl)azanium;molecular bromine;hydrobromide
C[N+](C)(C)CC1=CC=CC=C1.Br.BrBr
Quantity 5g
Melting Point 99°C to 101°C
Formula Weight 389.97
Percent Purity 98%
Chemical Name or Material Benzyltrimethylammonium tribromide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning