Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltriphenylphosphonium borohydride, 95%, Acros Organics

Click to view available options
Quantity:
5g
25g
18117-29-8
C25H22P·BH4
368.27
ICVMMQSETPKHRH-UHFFFAOYSA-N
benzyl triphenyl phosphanium;boron 1-,benzyltriphenylphosphonium tetrahydroborate
11303127
benzyl(triphenyl)phosphanium;boron(1-)
[B-].C1=CC=C(C=C1)C[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
18117-29-8
C25H22P·BH4
368.27
ICVMMQSETPKHRH-UHFFFAOYSA-N
benzyl triphenyl phosphanium;boron 1-,benzyltriphenylphosphonium tetrahydroborate
11303127
benzyl(triphenyl)phosphanium;boron(1-)
[B-].C1=CC=C(C=C1)C[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
Reduction reagent
Melting Point 157.0°C to 163.0°C
Quantity 25g
Assay Percent Range 95%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,dichloromethane,insoluble in benzene and hexane
Formula Weight 368.27
Percent Purity 95%
Chemical Name or Material Benzyltriphenylphosphonium borohydride

GHS Signal Word: Danger