Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylurea, 93%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
538-32-9
C8H10N2O
150.181
MFCD00007951
RJNJWHFSKNJCTB-UHFFFAOYSA-N
1-benzylurea,n-benzylurea,benzylcarbamide,urea, phenylmethyl,phenylmethylurea,urea, benzyl,cc-pmlsc-dma-p105,amino-n-benzylamide,benzyl urea,benzyl-urea
10853
benzylurea
C1=CC=C(C=C1)CNC(=O)N
538-32-9
C8H10N2O
150.181
MFCD00007951
RJNJWHFSKNJCTB-UHFFFAOYSA-N
1-benzylurea,n-benzylurea,benzylcarbamide,urea, phenylmethyl,phenylmethylurea,urea, benzyl,cc-pmlsc-dma-p105,amino-n-benzylamide,benzyl urea,benzyl-urea
10853
benzylurea
C1=CC=C(C=C1)CNC(=O)N
Quantity 25g
Melting Point 143°C to 151°C
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
CAS Min % 91.0
CAS Max % 100.0
Assay Percent Range 93%
Linear Formula C6H5CHNHC(O)nH
Merck Index 15, 1149
Formula Weight 150.18
Physical Form Powder
Percent Purity ≥91%
Chemical Name or Material Benzylurea, 93%
Show More Show Less