Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berberine Chloride, JP, 95-102%, Spectrum™

Click to view available options
Quantity:
5 kg
25 kg
Packaging:
Fiber Drum
Poly Pail
633-65-8
C20H18ClNO4
371.82
VKJGBAJNNALVAV-UHFFFAOYSA-M
16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(21),2,4(8),9,13,15,17,19-octaen-13-ylium chloride
[Cl-].COC1=CC=C2C=C3C4=CC5=C(OCO5)C=C4CC[N+]3=CC2=C1OC
633-65-8
C20H18ClNO4
371.82
VKJGBAJNNALVAV-UHFFFAOYSA-M
16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(21),2,4(8),9,13,15,17,19-octaen-13-ylium chloride
[Cl-].COC1=CC=C2C=C3C4=CC5=C(OCO5)C=C4CC[N+]3=CC2=C1OC

B2310, 633-65-8, C20H18NO4Cl

Spectrum™ Berberine Chloride, JP - Spectrum Chemical JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Quantity 25 kg
Ignition Residue 0.001
Water 8-12%
CAS Max % 1
Percent Purity 95 to 102%