Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bergamot Oil, Natural, Spectrum™

Click to view available options
Quantity:
25 mL
100 mL
500 mL
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
8007-75-8
8007-75-8

BE161, 8007-75-8

Spectrum™ Bergamot Oil, Natural , also known as Bergamot essential oil, is used as a top note in several perfumes. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Poly Pail
Quantity 5 kg
CAS Max % 100%
UN Number UN1169