Learn More
Siemens Healthineers Berichrom Heparin UF Controls
For use as an assayed control for the quantitative measurement of unfractionated heparin with Berichrom Heparin.
$207.00
Specifications
Type | Coagulation Control |
---|---|
For Use With (Application) | Heparin Testing |
Quantity | 6 x 1 mL |
Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
23-044-671
|
Siemens Healthineers
10445985 |
Low |
Pack of 6 for $207.00
|
|
|||||
23-044-672
|
Siemens Healthineers
10445986 |
High |
Pack of 6 for $207.00
|
|
|||||
Description
Berichrom Heparin UF Controls are high-level assayed controls that are used to monitor the performance of unfractionated heparin with the Berichrom Heparin assay.Specifications
Coagulation Control | |
Heparin Testing | |
6 x 1 mL |