Learn More
Berkshire™ BCR™ Cleanroom Bond 850 Paper
Manufacturer:  Berkshire™ BB85.0811.10C3/MRK
 View more versions of this product
Catalog No. 19-041-266
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Cellulose
Yellow (10/Pk.)
For photocopying or offset printing, or as lot tracer sheets, batch records or standard pages in cleanroom manuals
8.5 in.
24 lb.
11 in.
Class A, B and C/ISO Class 3-7
Yes
8.5 x 11 in. (216 x 279mm)
Standard photocopiers and laser printers
216mm
10.8kg
279mm
Cleanroom Paper