Learn More
Please call 1-800-766-7000 to place your order or try our site again later.
Berkshire BCR Cleanroom Bond 850 Paper
$18.00 - $28.00
Specifications
Dimensions (L x W) | 8.5 x 11 in. (216 x 279mm) |
---|---|
For Use With (Application) | For photocopying or offset printing, or as lot tracer sheets, batch records or standard pages in cleanroom manuals |
For Use With (Equipment) | Standard photocopiers and laser printers |
Length (English) | 8.5 in. |
Material | Cellulose |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
19-041-259
|
Berkshire
BB85081110W |
White |
Pack of 250 for $18.00
Case of 10 PK for $180.00 |
N/A |
19-041-260
|
Berkshire
BB85081110B |
Blue |
Pack of 250 for $19.00
Case of 10 PK for $190.00 |
N/A |
19-041-261
|
Berkshire
BB85081110C |
Yellow |
Pack of 250 for $19.00
Case of 10 PK for $190.00 |
N/A |
19-041-263
|
Berkshire
BB85081110G |
Green |
Pack of 250 for $19.00
Case of 10 PK for $190.00 |
N/A |
19-041-262
|
Berkshire
BB85081110P |
Pink |
Pack of 250 for $19.00
Case of 10 PK for $190.00 |
N/A |
19-041-266
|
Berkshire
BB85.0811.10C3/MRK |
Yellow (10/Pk.) |
Pack of 250 for $28.00
Case of 10 PK for $280.00 |
N/A |
Description
Heavier than the 680 series paper. Durable for double-sided documents, yet economical. Available in white, pink, yellow, blue, and green 8.5 x 11 in. (22 x 28cm) sheets.
Designed for cleanroom environments, BCR bond is made with the purest quality cellulose from specially selected forests and is specially coated to minimize particle and fiber release.
- Use these ultraclean, heat-resistant pages in any class of cleanroom for photocopying or offset printing, or as lot tracer sheets, batch records, or standard pages in cleanroom manuals
- Compatible with all standard photocopiers and laser printers without jamming
- Fine cut edges reduce particle contamination
- Low static charge prevents pages from sticking together
- Double-wrapped to ensure optimum cleanliness
- Autoclavable
Order Info
250 sheets per pack. Paper weight, 24 lb. (85.5g/m2).
Specifications
8.5 x 11 in. (216 x 279mm) | |
Standard photocopiers and laser printers | |
Cellulose | |
11 in. |
For photocopying or offset printing, or as lot tracer sheets, batch records or standard pages in cleanroom manuals | |
8.5 in. | |
Cleanroom Paper |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.