Learn More
Berkshire™ BCR™ Cleanroom Mopping System: Mop 4
Looped-end construction and fantail design minimize particle generation
Manufacturer:  Berkshire™ BCM.4S.12
 View more versions of this product
Catalog No. 19-121-717
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
White
Yes
BCR Mop 4 Sterile
Easy removal of contamination from floors and for the distribution of disinfectants
11.81 in.
ISO Class 4
Knitted Polyester
Yes
20 in.