Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Berkshire™ BCR™ Polyester Half-Finger Cleanroom Glove Liners
Continuous filament knitted polyester construction is thin yet durable, while the half-finger design permits tactility in addition to wicking moisture from the palms.
Supplier: Berkshire™ BGL2.200BR
This item is not returnable.
View return policy
Description
- Reduces perspiration and contact dermatitis commonly associated with wearing latex, urethane, PVC, or nitrile gloves
- Half-finger design permits tactility while also wicking moisture from the palms
- 100% continuous filament knitted polyester is thin, yet durable
- Ambidextrous design can be used on either hand
- Gloves can be repeatedly laundered for reuse
- Suitable for use in ISO Class Cleanrooms
Specifications
Uncoated | |
Reusable | |
Polyester | |
Half | |
Knit | |
Case |
White | |
Ambidextrous | |
Regular | |
Laboratories and Manufacturing Environments | |
White | |
BGL2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction