Berkshire™ Durx™ 770 Wipers

For ISO Class 5 (Class 100) applications

Manufacturer: Berkshire™ DR770.1212.20

 View more versions of this product

Catalog No. 17-987-801

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance RoHS
    Color White
    Compatibility Common cleaning and disinfecting solutions
    Dimensions (L x W) 12 x 12 in. (30.48 x 30.48cm)
    For Use With (Equipment) Cleaning of lab equipment, integrated circuit, printed circuit board and electronic components
    Material Polyester, Cellulose
    Width (English) 12 in.
    Width (Metric) 30.48cm
    Length (English) 12 in.
    Length (Metric) 30.48cm
    Product Type Durx™ 770 cleanroom wipes
    Autoclavable Yes
    Basis Weight 65.8
    For Use With (Application) For use in ISO Class 5 and higher cleanroom environments, facilities manufacturing integrated circuit, printed circuit board and electronic components

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit