Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Berkshire Fastsorb 820 Wipers
For use in ISO Class 5 (Class 100) applications
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
19-037260
|
Berkshire
FS820 0707 20 |
N/A | Size: 7 x 7 in. |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-037-601
|
Berkshire
FS820.0404.45 |
N/A | Size: 4 x 4 in. |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-037261
|
Berkshire
FS820 0909 16 |
N/A | Size: 9 x 9 in |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- Made of hydroentangled 70% rayon/30% polyester nonwoven that absorbs 7.5 times its weight
- Extremely soft, absorbent, and nonabrasive wiper wiping abrasion-sensitive surfaces and controlling spills
- Cushion-like with lofty caliper for packaging optics and other sensitive products
- Works well with IPA and other solvents
- Low fiber and particle generation
- Gentle enough for wiping and polishing soft metal and glass surfaces
Order Info
Basis weight, 77.6g/m2.
Specifications
Polyester, Rayon |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title