Learn More
Berkshire™ Knitted Polyester Lab-Tips Swabs
For cleaning grooved, slotted and recessed areas
$26.55 - $263.00
Specifications
Certifications/Compliance ISO Class 4
Handle Color Green
Product Type Swab
Tip Material Polyester
View More Specs
Products 4
Catalog Number Mfr. No. Length (Metric) Handle Length (Metric) Tip Tip Style Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Handle Length (Metric) Tip Tip Style Price Quantity & Availability  
19-041-364
View Documents
Berkshire™
LTP70R.20
58mm 12.5mm Rigid
Pack of 100 for $27.25
Case of 20 PK for $515.00
Add to Cart
19-041-363
View Documents
Berkshire™
LTP70F.20
59mm 10.9mm Flexible
Pack of 100 for $26.55
Case of 20 PK for $505.50
Add to Cart
19-041-368
View Documents
Berkshire™
LTP1465.10
144mm 16.8mm Flexible
Pack of 100 for $60.50
Case of 10 PK for $575.00
Add to Cart
19-041-370
View Documents
Berkshire™
LTP125.5
100mm 2.5mm Rigid
Case of 500 for $263.00
Add to Cart
Description
Description
  • Continuous filament polyester knit head
  • Short polypropylene handle with a rigid head
  • Thermally bonded head
  • Laundered and packaged in Class 10 (ISO 4) cleanroom
  • Autoclavable
Specifications
Specifications
ISO Class 4
Green
Swab
Polyester