Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berkshire™ Knitted Polyester Lab-Tips Swabs

For cleaning grooved, slotted and recessed areas

Manufacturer:  Berkshire™ LTP70R.20

 View more versions of this product

Catalog No. 19-041-364



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

2.8 in.
ISO Class 4
2.8mm
Green
0.49 in.
12.5mm
3.8mm
Rigid
3.8mm
Swab
0.11 in.
Designed for the highest level of contamination control in critical processing applications, cleaning of sensitive equipment, applying lubricants and other liquids, removing excess materials
2.28 in.
58mm
70mm
Polyester
0.15 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit