Learn More
Berkshire™ Pro-Wipe™ 750 Wipers
SDP
For ISO Class 5 (Class 100) applications
Supplier:  Berkshire™ PW750.0909.20
 View more versions of this product
Catalog No. 18-999-2564A
Add to cart
This item is not returnable. View return policy
Description
Description
  • Inner core of cellulose ultrasonically bonded to outer layers of polypropylene
  • Strong, clean, economical wiper is excellent for all general-purpose applications
  • Solvent resistant and super absorbent
Order Info
Basis weight: 71.5g/m2.
Certifications
Certified for contact with food per EC regulation 1935 (2004)
Specifications
Specifications
Pro-Wipe™ 750 Cleanroom Wipes
22.86 cm
22.86 cm
Super
71.5
9 x 9 in. (22.86 x 22.86 cm)
For tray lining and other lab applications, cleaning scratch sensitive surfaces
9 in.
9 in.
Cellulose, Polypropylene
Autoclavable
EC regulation 1935 (2004)
For ISO Class 5 (Class 100) applications, general spill pick up and cleaning surfaces, removal of etchants, acids, and other chemical spills