Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BERP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540957
Description
Peptide sequence: TICGAKQKVL QSQLDTLRQG QEHIGSSCSF AEQALRLGSA PEVLLVRKHM Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%; Zebrafish: 92%.
The protein encoded by this gene is a member of the tripartite motif family, also called the 'RING-B-box-coiled-coil' subgroup of RING finger proteins. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic filaments. It is similar to a rat protein which is a specific partner for the tail domain of myosin V, a class of myosins which are involved in the targeted transport of organelles. The rat protein can also interact with alpha-actinin-4. Thus it is suggested that this human protein may play a role in myosin V-mediated cargo transport. Alternatively spliced transcript variants encoding the same isoform have been identified.
Specifications
BERP | |
Polyclonal | |
Unconjugated | |
TRIM3 | |
Berp; BERP1; brain expressed ring finger; brain-expressed RING finger protein; Hac1; RING finger protein 22; RING finger protein 97; RING finger protein HAC1; RNF22; RNF97; Trim3; tripartite motif containing 3; tripartite motif protein 3; tripartite motif protein TRIM3; tripartite motif-containing 3; tripartite motif-containing protein 3 | |
Rabbit | |
Protein A | |
RUO | |
10612 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75382 | |
TRIM3 | |
Synthetic peptide directed towards the N-terminal of human TRIM3 (aa 225-274). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction