Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beryllium 2,4-pentanedionate, White powder, 99%, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
10210-64-7
C10H14BeO4
207.23
MFCD00013485
BBKXDHBLPBKCFR-OHSCNOFNSA-L
beryllium bis 4-oxo-2-pentene-2-olate
101644356
beryllium;(Z)-4-oxopent-2-en-2-olate;(E)-4-oxopent-2-en-2-olate
[Be+2].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-]
10210-64-7
C10H14BeO4
207.23
MFCD00013485
BBKXDHBLPBKCFR-OHSCNOFNSA-L
beryllium bis 4-oxo-2-pentene-2-olate
101644356
beryllium;(Z)-4-oxopent-2-en-2-olate;(E)-4-oxopent-2-en-2-olate
[Be+2].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-]
Assay Percent Range 99%
Formula Weight 207.23
Physical Form Powder
Odor Odorless
Color White
Quantity 100g
Chemical Name or Material Beryllium 2, 4-pentanedionate

H301-H330-H315-H319-H317-H335-H350-H372-H411: Toxic if swallowed. Fatal if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packaging Group: II

EINECSNumber : 233-513-5