Beryllium 2,4-pentanedionate, White powder, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar 3857514

Quantity 25g

 View more versions of this product

Catalog No. AA3857514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10210-64-7
    Physical Form Powder
    Assay Percent Range 99%
    Molecular Formula C10H14BeO4
    Formula Weight 207.23
    MDL Number MFCD00013485
    Synonym Beryllium acetylacetonate
    Chemical Name or Material Beryllium 2, 4-pentanedionate

    Specifications

    Physical Form Powder
    Formula Weight 207.23
    Quantity 25g
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: II
    EINECS Number 233-513-5
    Assay Percent Range 99%
    Molecular Formula C10H14BeO4
    MDL Number MFCD00013485
    CAS 10210-64-7
    Odor Odorless
    Synonym Beryllium acetylacetonate
    Health Hazard 1 H301-H330-H315-H319-H317-H335-H350-H372-H411: Toxic if swallowed. Fatal if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.
    Chemical Name or Material Beryllium 2, 4-pentanedionate

    H301-H330-H315-H319-H317-H335-H350-H372-H411: Toxic if swallowed. Fatal if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit