Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Beryllium foil, 0.5mm (0.02 in.) thick, 99.4% (metals basis)

Click to view available options
Quantity:
25 x 25mm
50 x 50mm
7440-41-7
Be
9.012
MFCD00134032
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
5460467
CHEBI:33783
beryllium
[Be]
7440-41-7
Be
9.012
MFCD00134032
ATBAMAFKBVZNFJ-UHFFFAOYSA-N
glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
5460467
CHEBI:33783
beryllium
[Be]
Quantity 25 x 25mm
Weight ≈0.6g/25x25mm
Odor Odorless
Physical Form Foil
Percent Purity 99.4%
Chemical Name or Material Beryllium foil, 0.5mm (0.02 in.) thick, Vacuum tight
Assay (metals basis)

GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P501c

H301-H315-H317-H319-H330-H335-H350i-H372

EINECSNumber : 231-150-7

TSCA : Yes

Recommended Storage : Ambient temperatures