Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beryllium ICP Standard, 1mL = 1mg Be (1,000ppm Be), Ricca Chemical

Click to view available options
Quantity:
100mL
500mL
Packaging:
Poly natural
19049-40-2,7697-37-2,19049-40-2
19049-40-2,7697-37-2,19049-40-2
CAS Min % 91.0,6.01,1.03
CAS Max % 95.0,6.26,1.07
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 1,000ppm Be
Equivalence 1 mL = 1 mg Be
Color Colorless
Quantity 500mL
Chemical Name or Material Beryllium ICP Standard