Beryllium plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar 13848AP

Quantity 500mL

 View more versions of this product

Catalog No. AA13848AP

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7697-37-2,19049-40-2,7732-18-5
    Physical Form Liquid
    Molecular Formula Be4O(C2H3O2)6 in 5% HNO3
    MDL Number MFCD00134032
    Chemical Name or Material Beryllium

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 500mL
    DOT Information Hazard Class: 8, Packing Group: III
    Molecular Formula Be4O(C2H3O2)6 in 5% HNO3
    CAS 7697-37-2,19049-40-2,7732-18-5
    Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Beryllium oxide acetate: 1.13%; Water: 93.87%
    Appearance Clear liquid
    MDL Number MFCD00134032
    Health Hazard 1 H350-H373-H314-H317: May cause cancer. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause an allergic skin reaction.
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Beryllium

    H350-H373-H314-H317: May cause cancer. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause an allergic skin reaction.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit