Alfa Aesar™ Beryllium, plasma standard solution, Specpure™, Be 10,000µg/mL

Manufacturer: Alfa Aesar™ 14406AP

Quantity 500mL

 View more versions of this product

Catalog No. AA14406AP

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00134032
    UN Number UN2922
    Chemical Name or Material Beryllium plasma standard solution, Be 10000μg/mL
    Grade Specpure™

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Concentration Be4O(C2H3O2)6 in 5% HNO3
    UN Number UN2922
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H350-H372-H314-H302-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Quantity 500mL
    Physical Form Liquid
    MDL Number MFCD00134032
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
    Chemical Name or Material Beryllium plasma standard solution, Be 10000μg/mL
    Grade Specpure™

    GHS H Statement
    H350-H372-H314-H302-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit