Beryllium Powder, -325 mesh, 99+% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1010822

Quantity 100g

 View more versions of this product

Catalog No. AA1010822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-41-7
    Physical Form Powder
    Assay Percent Range 99+% (Metals basis)
    MDL Number MFCD00134032
    Chemical Name or Material Beryllium

    Specifications

    Quantity 100g
    Color Gray
    DOT Information DOT Class: 6.1, Packaging Group: II
    EINECS Number 231-150-7
    Physical Form Powder
    Assay Percent Range 99+% (Metals basis)
    MDL Number MFCD00134032
    CAS 7440-41-7
    Odor Odorless
    Health Hazard 1 H301-H330-H315-H319-H317-H335-H350-H372; Toxic if swallowed. Fatal if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. May cause cancer. Causes damage to organs through prolonged or repeated exposure.
    Appearance Gray powder
    Chemical Name or Material Beryllium

    H301-H330-H315-H319-H317-H335-H350-H372; Toxic if swallowed. Fatal if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. May cause cancer. Causes damage to organs through prolonged or repeated exposure.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit