Alfa Aesar™ Beryllium Thinfoil, 0.01mm (0.0004 in.) thick, 99.8% (metals basis)

Manufacturer: Alfa Aesar™ 41612FF

Quantity 25x25mm

 View more versions of this product

Catalog No. AA41612FF

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-41-7
    MDL Number MFCD00134032
    Chemical Name or Material Beryllium Thinfoil, 0.01mm (0.0004 in.) thick, Not certified pinhole free

    Specifications

    Health Hazard 2 GHS H Statement
    H301-H330-H350-H372-H315-H319-H317-H335
    Toxic if swallowed.
    Fatal if inhaled.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Foil
    MDL Number MFCD00134032
    CAS 7440-41-7
    Odor Odorless
    Health Hazard 1 Danger
    Weight ≈0.012g/25x25mm
    Quantity 25x25mm
    Chemical Name or Material Beryllium Thinfoil, 0.01mm (0.0004 in.) thick, Not certified pinhole free
    Assay 99.8%, (metals basis)

    GHS H Statement
    H301-H330-H350-H372-H315-H319-H317-H335
    Toxic if swallowed.
    Fatal if inhaled.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit