Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Beryllium wire, 0.125mm (0.005 in.) dia., annealed, 99.7% (metals basis)

Manufacturer:  Alfa Aesar™ 42471G1

Quantity 1m

 View more versions of this product

Catalog No. AA42471G1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-41-7
MFCD00134032
Beryllium wire, 0.125mm (0.005 in.) dia., Annealed, Temper: soft

Specifications

≈0.023g/m
7440-41-7
MFCD00134032
Beryllium wire, 0.125mm (0.005 in.) dia., Annealed, Temper: soft
1m
Wire
Odorless
99.7%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit