SHOWA™ Answer™ and Answer Plus™ Gloves

Combine the look and feel of leather with the durability and flexibility of a nitrile-coated fabric

Manufacturer: SHOWA™ P273507

 View more versions of this product

Catalog No. 11-395-7B

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Slip-on
    For Use With (Application) assembly, fabrication, maintenance, inspection, automotive assembly, material handling
    Cut Resistance Rating Level 1
    Cleaning Procedure Launder at 40°C using a neutral Detergent
    Exterior Finish Textured
    Length (English) 8 in.
    Length (Metric) 20.32cm
    Material Nitrile
    Liner Material Cotton Jersey, Polyester
    Powdered No
    Size 7
    Coated Area Full
    Color Navy

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit