Learn More
SHOWA™ Answer™ and Answer Plus™ Gloves
Combine the look and feel of leather with the durability and flexibility of a nitrile-coated fabric
Manufacturer:  SHOWA™ P273509
 View more versions of this product
Catalog No. 11-395-7D
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Both styles feature special nitrile compound laminated onto exterior of cotton-poly base; the result is extra strength
  • Unique patterned finish provides excellent wet and dry grip
  • Sanitized™ interior, an antimicrobial, bacteriostatic treatment that does not wash out with laundering, extends the life of the gloves
  • Perforated back of Answer Plus version keeps hands extra cool
  • Talc-free to avoid silica contamination
  • Launderable
Compliance
Compliant with 21CFR170-199.
Specifications
Specifications
Safety
Large
23 cm
Nitrile
9 in.