Learn More
SHOWA™ D-Flex™ Plus Gloves
Similar to D-Flex gloves (above), but ideal for medium-duty applications
Manufacturer:  SHOWA™ 91708
 View more versions of this product
Catalog No. 19-048-579C
Add to cart
Description
Description
  • Dotted style comes in hand-specific versions
  • Medium weight
  • Launderable
  • 7-gauge
  • Made in the U.S.A.
Specifications
Specifications
Safety
Ambidextrous
Food processing, machine or compartment assembly, bottling, sheet Metal handling, construction, recycling, Glass handling, razor blades
8
White
Level 5
Blue
HPPE
Medium weight