Learn More
SHOWA™ D-Flex™ Plus Gloves
Click to view available options
Size:
6
7
8
9
10
Hand:
Ambidextrous
Left
Right
Coating Material:
PVC Dots
Uncoated
Description
- Dotted style comes in hand-specific versions
- Medium weight
- Launderable
- 7-gauge
- Made in the U.S.A.
Specifications
Coating Material | PVC Dots |
Cut Resistance Rating | ANSI A5 |
Hand | Right |
Material | HPPE |
Product Type | Cut-resistant Gloves |
Size | 10 |