SHOWA™ DermaThin™ Natural Rubber Gloves

Thin gauge disposable gloves that offer outstanding strength, wear and dexterity, and are cooler, more durable, better fitting

Manufacturer: SHOWA™ W1005M

 View more versions of this product

Catalog No. 11-394-17A

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Glove Cuff Style Rolled
    For Use With (Application) cosmetic, electronic assembly, pharmaceutical, food processing applications
    Length (English) 9.5 in.
    Length (Metric) 24cm
    Material Latex
    Thickness 5mil
    Size Medium
    Color White
    Powdered Yes
    Coating Material Smooth

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit