SHOWA™ Bex™ Sof Paw Gloves

Provide excellent abrasion resistance and good grip, wet or dry

Manufacturer: SHOWA™ 7007

 View more versions of this product

Catalog No. 19-014571

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Slip-on
    Cleaning Procedure Washable
    Exterior Finish Textured
    For Use With (Application) assembly, inspection, material handling, maintenance
    Material PVC
    Liner Material Cotton
    Size 7
    Sterile Yes
    Coated Area Full
    Color Yellow
    Cut Resistance Rating Level 1

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit