Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Char-Guard Gloves

Manufacturer:  SHOWA™ 881410

 View more versions of this product

Catalog No. 19-150-1282


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Textured
handling hot objects, bakeries, machine assembly, shipping and receiving, food service, autoclave, inspection, landscaping
X-Large
260°C
Black
35.56cm
Launder at 40°C using a neutral Detergent
Thermal Fabric
500°F
CFIA, 21 CFR
14 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit