Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Hustler™ Gloves

Provides excellent abrasion resistance and exceptional protection against solvents and acids

Manufacturer:  SHOWA™ 728R10

 View more versions of this product

Catalog No. 11-388-132



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Rough
Petrochemical, refining and general chemical applications, including Acids, Oils, Grease, Caustics and many Solvents
Nitrile,PVC (Polyvinyl Chloride)
21mil
Red
45.7cm
Gauntlet
Cotton interlock
10
CE
18 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit