Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ Nitri-Dex™ Flock-Lined Nitrile Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
Unsupported, flock-lined nitrile coated gloves feature a lasting grip and provide protection from oils, hydrocarbons, grease and abrasion.
- Ideal for jobs requiring comfort, tactility, and contact with chemical hazards
- Flocked cotton liner ensures easy donning and doffing
- Rolled cuff prevents debris from entering
- Lightweight nitrile material helps reduce hand fatigue
Specifications
ANSI Abrasion Rating | ANSI 1 |
Certifications/Compliance | 21 CFR 170-199 |
Certifications/Compliance (Europe) | CE |
Chlorinated | No |
Color | Blue |
Latex Free | Yes |
Material | Nitrile |
Size | X-Large |
Thickness (English) | 9 mil |
Thickness (Metric) | 0.23 mm |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction