SHOWA™ Glove™ Nitri-Flex™ Lite and Nitri-Flex Plus Nitrile-Coated Gloves

Manufacturer: SHOWA™ 460008

 View all options for this product

Catalog No. 19-050-284B

  Check Availability
Add to Cart


    Coated Area Palm
    Color Light green, green
    Cut Resistance Rating Level 1
    Cleaning Procedure Launder at 40°C using a neutral Detergent
    Exterior Finish Textured
    Glove Cuff Style Knit
    For Use With (Application) small parts assembly, inspection, shipping and receiving, light fabrication, material handling, automotive, packaging
    Length (English) 9 in.
    Length (Metric) 23cm
    Material Nitrile
    Liner Material Cotton
    Size 8

    Nitri-Flex™ Lite

    • Flat-dipped nitrile, seamless nylon liner
    • Smooth palm
    • Light green with green dip
    Nitri-Flex™ Plus
    • Flat dipped nitrile, seamless cotton liner
    • Provides superior absorption of perspiration
    • Textured palm provides excellent wet, dry, or oily grip

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit