SHOWA™ Glove™ Nitri-Solve™ Unlined Nitrile Gloves
Manufacturer: SHOWA™ 72708
 View more versions of this product
Catalog No. 11-395-30B
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR 170-199, NFPA 1992, CFIA, CE
    Glove Cuff Style Straight
    For Use With (Application) manufacturing, chemical processing, Petrochemicals, food processing, aerospace, automobile degreasing, automotive assembly, painting applications, machine applications using cutting Oils
    Length (English) 13 in.
    Length (Metric) 33cm
    Material Nitrile
    Size Medium
    Thickness 15mil
    Coating Material Bisque
    Color Light green
  • Description & Specifications
Related Products
Product Content Feedback