Learn More
SHOWA™ SHOWA™ Industrial Unlined Nitrile Green Gloves, 12-pack
Wear SHOWA™ Industrial Unlined Nitrile Green Gloves for applications in manufacturing, chemical processing, Petrochemicals, food processing, aerospace, automobile degreasing, automotive assembly, painting applications and more.
Manufacturer:  SHOWA™ 72709
 View more versions of this product
Catalog No. 11-395-30C
Add to cart
Description
Description
  • Cat III
  • 12-pack
  • Case-hardened finish enhances chemical and abrasion resistance
  • Easy to pull on and off
  • TAA compliant
Specifications
Specifications
Safety
manufacturing, chemical processing, Petrochemicals, food processing, aerospace, automobile degreasing, automotive assembly, painting applications, machine applications using cutting Oils
Large
Light Green
33 cm
Straight
Nitrile
15mil
13 in.
21 CFR 170-199, CFIA, CE