Learn More
SHOWA™ SHOWA™ Industrial Unlined Nitrile Green Gloves, 12-pack
Wear SHOWA™ Industrial Unlined Nitrile Green Gloves for applications in manufacturing, chemical processing, Petrochemicals, food processing, aerospace, automobile degreasing, automotive assembly, painting applications and more.
Manufacturer:  SHOWA™ 72710
 View more versions of this product
Catalog No. 11-395-30D
Add to cart
Description
Description
  • Cat III
  • 12-pack
  • Case-hardened finish enhances chemical and abrasion resistance
  • Easy to pull on and off
  • TAA compliant
Specifications
Specifications
Safety
manufacturing, chemical processing, Petrochemicals, food processing, aerospace, automobile degreasing, automotive assembly, painting applications, machine applications using cutting Oils
X-Large
Light Green
33 cm
Straight
Nitrile
15mil
13 in.
21 CFR 170-199, CFIA, CE