Learn More
SHOWA™ SHOWA™ Industrial Unlined Nitrile Green Gloves, 12-pack
Wear SHOWA™ Industrial Unlined Nitrile Green Gloves for applications in manufacturing, chemical processing, Petrochemicals, food processing, aerospace, automobile degreasing, automotive assembly, painting applications and more.
Manufacturer:  SHOWA™ 74710
 View more versions of this product
Catalog No. 11-395-32B
  Check Availability
Add to cart
Specifications
Description
  • Cat III
  • 12-pack
  • Case-hardened finish enhances chemical and abrasion resistance
  • Easy to pull on and off
  • TAA compliant
Description & Specifications
Specifications
Bisque
manufacturing, chemical processing, Petrochemicals, food processing, aerospace, automobile degreasing, automotive assembly, painting applications, machine applications using cutting Oils
X-Large
21 CFR 170-199, NFPA 1992, CFIA, CE
19 in.
Straight
Nitrile
22mil
Light Green
48.26cm