Learn More
SHOWA™ Nitty Gritty™ Gloves
Cut resistant and recyclable
Manufacturer:  SHOWA™ L63PNFW08
 View more versions of this product
Catalog No. 19-014-953
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Palm
Launder at 40°C using a neutral Detergent
Knit
Rubber, natural
8
Level 2
Wrinkle
Glass handling, bottling, lumber, brick and tile handling, maintenance
Cotton
White,Yellow