SHOWA™ Nitty Gritty™ Gloves

Cut resistant and recyclable

Manufacturer: SHOWA™ 63NFW10

 View more versions of this product

Catalog No. 19-014-934

  Check Availability
Add to Cart

    Specifications

    Exterior Finish Wrinkle
    Glove Cuff Style Gauntlet
    For Use With (Application) Glass handling, bottling, lumber, brick and tile handling, maintenance
    Liner Material Cotton
    Cut Resistance Rating Level 2
    Cleaning Procedure Launder at 40°C using a neutral Detergent
    Material Natural Rubber
    Size 10
    Coated Area Full
    Color Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit