Learn More
SHOWA™ Terry-Plus™ Gloves
Added air cushion insulates against moderately high temperatures
Manufacturer:  SHOWA™ 937C10
 View more versions of this product
Catalog No. 19-048-581B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Heavyweight 24 oz. construction makes these seamless knit, Kevlar™/cotton terrycloth ambidextrous gloves suitable for a variety of fabricating and material handling applications.
  • High Kevlar/cotton loops provide additional cut resistance
  • Loops also form an air cushion that protects against moderate temperatures (up to 177°C [350°F] for intermittent contact)
  • Dots on both front and back of gloves provide better gripping
  • Color coded with an overcasting on the knitwrist for easy size identification
  • Launderable
Not for use with moving or serrated blades.
Specifications
Specifications
Safety
Knit
10
177°C
24 oz.
Neoprene
Dupont™ Kevlar™ Aramid Fiber, Terrycloth
350°F
680.38 g
Black