Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Seamless Knit Cut-Resistant Gloves, PVC Dot Grip

Catalog Number p-144254
Click to view available options
Size:
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

High loops of aramid fiber and cotton provide air cushion and insulation. PVC dots on both sides of the glove surface increase grip capabilities and tactile precision.

  • Seamless knit construction reduces irritation and chaffing
  • PVC dots on both sides of the glove for enhanced grip capabilities
  • Elasticated wrist helps prevent dirt and debris from entering the glove
  • Thermal insulation
  • Machine washable
Cuff Material Elastic
Coating Material PVC Dots (2 Sides)
Color Yellow
Cut Resistance Rating ANSI A3
Material HPPE
Product Type Cut-resistant Gloves
Size 9
For Use With (Application) Automotive Maintenance/Repairs, Heavy Equipment Handling, HVAC, and Metal Stamping
Glove Cuff Color Natural
Glove Cuff Style Knit
Series 937C
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit