SHOWA™ Viton™ and Viton II Gloves

Provide highest chemical-resistance barrier to aromatic hydrocarbons such as benzene, toluene, or xylene.

Manufacturer: SHOWA™ 89009

 View more versions of this product

Catalog No. 19-052-732

  Check Availability
Add to Cart

    Specifications

    Certifications CE
    Exterior Finish Smooth
    Glove Cuff Style Rolled
    For Use With (Application) Provides high temperature and resistance to fuels used in the aerospace industry
    Length (English) 14 in.
    Length (Metric) 35.5cm
    Material Viton™, Butyl
    Size 9
    Thickness 28mil
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit