Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Zorb-IT Ultra Work Gloves

Catalog Number p-3766654
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Work gloves ideal for medium to heavy duty tasks in a variety of industries.

Color Gray, Yellow
Cut Resistance Rating ANSI A3
Material Aramid Fiber, Lycra
Product Type Cut-resistant Gloves
Size 7
For Use With (Application) Automotive, construction, glass, metallurgy
Glove Cuff Style Elasticated
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit