Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ N-DEX™ 7005 Nitrile Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Description
- 100% nitrile, latex-free
- Powder free
- Increased dexterity
- 4 mil thick, 9.5 in. length
- Protects hands from an array of chemical hazards
- Rolled cuff prevents debris from entering the glove
- Second-skin feel reduces hand fatigue
- Ideal for a variety of applications including blood drawing, dental hygiene, food preparation, EMS, lab work and pharmaceutical
- Made in the USA
Compliance
Data Testing is available upon request.
Specifications
Certifications/Compliance | 21 CFR 170-199, FDA, NFPA, OSHA, CFIA, ASTM D5526 |
Applications | Laboratory, Healthcare, Industrial |
AQL | 1.5 |
Color | Blue |
Disposable | Single-use |
Exterior Finish | No Texture |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Size | X-Small |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction