SHOWA™ Natural Rubber Latex HD™ Gloves
Two-color wear indicator
Manufacturer: SHOWA™ 55809
 View more versions of this product
Catalog No. 19-019887
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR 170-199, EN 388 1120, CE
    Exterior Finish Bisque
    Glove Cuff Style Rolled
    For Use With (Application) manufacturing, aircraft assembly, janitorial, plant maintenance, agriculture, heavy duty plant maintenance, chemical processing, sanitization clean up, sandblasting/ electroplating
    Length (English) 18 in.
    Length (Metric) 45.7cm
    Material Natural Rubber
    Powdered Yes
    Size Large
    Sterile No
    Thickness 40mil
    Color Black, red
  • Description & Specifications
Related Products
Product Content Feedback